Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajc17
Entrez Gene ID
311329
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 17
Gene Aliases
RGD1308015
Chromosome Location
Chr.3: 110980074 - 111013602 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191740.1 NP_001178669.1 7 - 8 549 75

View Full Product Details