Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss58
Entrez Gene ID
408204
Gene Name
protease, serine, 58
Gene Aliases
Tryx3, try1
Chromosome Location
Chr.4: 69145521 - 69149477 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009174.1 NP_001009174.1 2 - 3 174 82

View Full Product Details