Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc9b1
Entrez Gene ID
365946
Gene Name
solute carrier family 9, subfamily B (NHA1, cation proton antiporter 1), member 1
Gene Aliases
Nhedc1, Nhedc2, RGD1305087
Chromosome Location
Chr.2: 240527130 - 240576179 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173973.1 NP_001167444.1 7 - 8 1174 86
XM_008761518.1 XP_008759740.1 6 - 7 690 86
GenBank mRNA BC158723.1 - 2 - 3 171 86

View Full Product Details