Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casp14
Entrez Gene ID
299587
Gene Name
caspase 14
Gene Aliases
-
Chromosome Location
Chr.7: 13938377 - 13944286 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241046.2 XP_006241108.1 1 - 2 174 65

View Full Product Details