Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Casp14
Entrez Gene ID
299587
Gene Name
caspase 14
Gene Aliases
-
Chromosome Location
Chr.7: 13938377 - 13944286 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241046.2 XP_006241108.1 2 - 3 234 71

View Full Product Details