Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem68
Entrez Gene ID
312946
Gene Name
transmembrane protein 68
Gene Aliases
RGD1309006
Chromosome Location
Chr.5: 16363911 - 16413307 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107903.1 NP_001101373.1 2 - 3 181 148

View Full Product Details