Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhdds
Entrez Gene ID
298541
Gene Name
dehydrodolichyl diphosphate synthase
Gene Aliases
-
Chromosome Location
Chr.5: 152200679 - 152227696 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011978.1 NP_001011978.1 7 - 8 785 66
XM_006239014.2 XP_006239076.1 7 - 8 900 66
GenBank mRNA BC085875.1 - 7 - 8 785 66

View Full Product Details