Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fktn
Entrez Gene ID
362520
Gene Name
fukutin
Gene Aliases
Fcmd
Chromosome Location
Chr.5: 70522001 - 70578270 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108667.1 NP_001102137.1 8 - 9 1375 69
XM_006238166.2 XP_006238228.1 10 - 11 1482 69
XM_006238168.2 XP_006238230.1 9 - 10 1409 69
XM_006238169.1 XP_006238231.1 8 - 9 1292 69
XM_008763712.1 XP_008761934.1 9 - 10 1502 69
XM_008763713.1 XP_008761935.1 10 - 11 1476 69

View Full Product Details