Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wfdc10
Entrez Gene ID
685216
Gene Name
WAP four-disulfide core domain 10
Gene Aliases
-
Chromosome Location
Chr.3: 161121690 - 161123043 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109461.1 NP_001102931.1 1 - 2 150 103

View Full Product Details