The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127297.1 | NP_001120769.1 | 9 - 10 | 1011 | 54 |
XM_008768366.1 | XP_008766588.1 | 9 - 10 | 1630 | 54 | |
XM_008768367.1 | XP_008766589.1 | 9 - 10 | 908 | 54 | |
GenBank mRNA | BC166829.1 | - | 9 - 10 | 1011 | 54 |