Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctns
Entrez Gene ID
287478
Gene Name
cystinosin, lysosomal cystine transporter
Gene Aliases
-
Chromosome Location
Chr.10: 59749721 - 59764819 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191647.1 NP_001178576.1 8 - 9 771 76

View Full Product Details