Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pde6g
Entrez Gene ID
688297
Gene Name
phosphodiesterase 6G, cGMP-specific, rod, gamma
Gene Aliases
-
Chromosome Location
Chr.10: 109620499 - 109626119 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750982.3 XP_003751030.1 1 - 2 1012 78

View Full Product Details