The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111366.1 | NP_001104836.1 | 2 - 3 | 181 | 78 |
XM_006247823.2 | XP_006247885.1 | 2 - 3 | 197 | 78 |