Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc48
Entrez Gene ID
287371
Gene Name
leucine rich repeat containing 48
Gene Aliases
RGD1309150
Chromosome Location
Chr.10: 46711518 - 46762261 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013857.1 NP_001013879.1 3 - 4 407 67
XM_006246462.2 XP_006246524.1 4 - 5 675 67
XM_006246463.2 XP_006246525.1 4 - 5 454 67
XM_006246464.2 XP_006246526.1 4 - 5 1006 67
XM_006246465.2 XP_006246527.1 4 - 5 567 67
XM_008767782.1 XP_008766004.1 5 - 6 561 67
XM_008767783.1 XP_008766005.1 4 - 5 1771 67
XM_008767784.1 XP_008766006.1 5 - 6 2047 67
XM_008767785.1 XP_008766007.1 4 - 5 454 67
GenBank mRNA BC083838.1 - 3 - 4 407 67

View Full Product Details