The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108289.1 | NP_001101759.1 | 4 - 5 | 445 | 93 |
XM_008768075.1 | XP_008766297.1 | 4 - 5 | 636 | 93 | |
GenBank mRNA | FQ213501.1 | - | 4 - 5 | 981 | 93 |
FQ216939.1 | - | 4 - 5 | 476 | 93 |