Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Maml1
Entrez Gene ID
303101
Gene Name
mastermind like 1 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.10: 35765712 - 35801375 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106997.1 NP_001100467.1 4 - 5 2329 82
XM_006246251.2 XP_006246313.1 4 - 5 3603 82

View Full Product Details