Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Socs7
Entrez Gene ID
287659
Gene Name
suppressor of cytokine signaling 7
Gene Aliases
-
Chromosome Location
Chr.10: 85301625 - 85337971 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006247484.2 XP_006247546.2 1 - 2 1033 99

View Full Product Details