Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Iba57
Entrez Gene ID
363611
Gene Name
IBA57, iron-sulfur cluster assembly homolog (S. cerevisiae)
Gene Aliases
RGD1306000
Chromosome Location
Chr.10: 45506124 - 45514909 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108827.1 NP_001102297.1 2 - 3 731 115

View Full Product Details