Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfp13
Entrez Gene ID
287095
Gene Name
zinc finger protein 13
Gene Aliases
-
Chromosome Location
Chr.10: 12908643 - 12916784 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105765.1 NP_001099235.1 4 - 5 702 57

View Full Product Details