Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmo1
Entrez Gene ID
360553
Gene Name
vitelline membrane outer layer 1 homolog (chicken)
Gene Aliases
RGD1559861
Chromosome Location
Chr.10: 57120499 - 57121584 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191823.1 NP_001178752.1 2 - 3 306 68

View Full Product Details