The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013033.2 | NP_037165.1 | 14 - 15 | 1949 | 81 |
GenBank mRNA | BC081827.1 | - | 14 - 15 | 1981 | 81 |
D16101.1 | - | 14 - 15 | 1949 | 81 | |
U03120.1 | - | 14 - 15 | 1907 | 81 |