Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hormad2
Entrez Gene ID
498400
Gene Name
HORMA domain containing 2
Gene Aliases
-
Chromosome Location
Chr.14: 84576857 - 84662239 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017501.1 NP_001017501.1 5 - 6 542 118
XM_008770340.1 XP_008768562.1 5 - 6 637 118
XM_008770341.1 XP_008768563.1 7 - 8 794 118
XM_008770342.1 XP_008768564.1 6 - 7 711 118
XM_008770343.1 XP_008768565.1 5 - 6 493 118
XM_008770344.1 XP_008768566.1 5 - 6 634 118
GenBank mRNA BC079253.1 - 5 - 6 542 118

View Full Product Details