Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpr64
Entrez Gene ID
266735
Gene Name
G protein-coupled receptor 64
Gene Aliases
He6, Re6
Chromosome Location
Chr.X: 36930186 - 37054925 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270871.1 NP_001257800.1 10 - 11 526 131
NM_001270872.1 NP_001257801.1 9 - 10 490 131
NM_181366.2 NP_852031.1 11 - 12 574 131
XM_006256916.2 XP_006256978.1 11 - 12 614 131
XM_008773154.1 XP_008771376.1 10 - 11 954 131
XM_008773155.1 XP_008771377.1 10 - 11 564 131
XM_008773156.1 XP_008771378.1 9 - 10 918 131
XM_008773157.1 XP_008771379.1 9 - 10 912 131
XM_008773158.1 XP_008771380.1 8 - 9 876 131
XM_008773159.1 XP_008771381.1 7 - 8 467 131
XM_008773160.1 XP_008771382.1 10 - 11 955 131
GenBank mRNA AF538953.1 - 11 - 12 584 131
AF538958.1 - 10 - 11 536 131
AF538959.1 - 9 - 10 500 131

View Full Product Details