Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mbtps2
- Entrez Gene ID
-
302705
- Gene Name
- membrane-bound transcription factor peptidase, site 2
- Gene Aliases
-
LRRGT00063
- Chromosome Location
-
Chr.X: 40258933 - 40309047 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC097391.1 |
- |
- |
725 |
65 |
View Full Product Details