Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Etv5
Entrez Gene ID
303828
Gene Name
ets variant 5
Gene Aliases
-
Chromosome Location
Chr.11: 82194657 - 82252145 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107082.1 NP_001100552.1 1 - 2 62 68

View Full Product Details