Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Insig2
Entrez Gene ID
288985
Gene Name
insulin induced gene 2
Gene Aliases
-
Chromosome Location
Chr.13: 37264818 - 37292730 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178091.4 NP_835192.1 4 - 5 659 93
XM_006249667.2 XP_006249729.1 5 - 6 1124 93
XM_006249668.2 XP_006249730.1 5 - 6 972 93
XM_006249669.2 XP_006249731.1 5 - 6 939 93
GenBank mRNA AY152392.1 - 4 - 5 659 93
BC085682.1 - 5 - 6 797 93

View Full Product Details