The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133589.2 | NP_598273.2 | 6 - 7 | 771 | 83 |
GenBank mRNA | BC078747.1 | - | 6 - 7 | 771 | 83 |
X77236.1 | - | 6 - 7 | 736 | 83 |