Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nkain4
Entrez Gene ID
296469
Gene Name
Na+/K+ transporting ATPase interacting 4
Gene Aliases
RGD1305809
Chromosome Location
Chr.3: 176444545 - 176465197 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006235744.2 XP_006235806.1 1 - 2 164 69

View Full Product Details