Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sugp2
Entrez Gene ID
361126
Gene Name
SURP and G patch domain containing 2
Gene Aliases
Sfrs14, Srsf14
Chromosome Location
Chr.16: 20908323 - 20939545 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108397.1 NP_001101867.1 1 - 2 415 100

View Full Product Details