Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Armc6
Entrez Gene ID
306352
Gene Name
armadillo repeat containing 6
Gene Aliases
RGD1306691
Chromosome Location
Chr.16: 20939311 - 20949732 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC166821.1 - - 802 81

View Full Product Details