Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csnk2a2
Entrez Gene ID
307641
Gene Name
casein kinase 2, alpha prime polypeptide
Gene Aliases
-
Chromosome Location
Chr.19: 9972537 - 10012043 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107409.1 NP_001100879.1 6 - 7 517 91
XM_006255082.1 XP_006255144.1 6 - 7 517 91
GenBank mRNA BC168200.1 - 6 - 7 533 91

View Full Product Details