Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Csnk2a2
Entrez Gene ID
307641
Gene Name
casein kinase 2, alpha prime polypeptide
Gene Aliases
-
Chromosome Location
Chr.19: 9972537 - 10012043 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107409.1 NP_001100879.1 7 - 8 619 78
XM_006255082.1 XP_006255144.1 7 - 8 619 78
GenBank mRNA BC168200.1 - 7 - 8 635 78

View Full Product Details