Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hepacam2
Entrez Gene ID
296846
Gene Name
HEPACAM family member 2
Gene Aliases
RGD1305697
Chromosome Location
Chr.4: 28403050 - 28437746 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106580.2 NP_001100050.1 3 - 4 741 117
XM_006236029.2 XP_006236091.1 3 - 4 794 117
XM_006236030.2 XP_006236092.1 3 - 4 823 117
GenBank mRNA BC168741.1 - 3 - 4 741 117

View Full Product Details