Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nceh1
Entrez Gene ID
294930
Gene Name
neutral cholesterol ester hydrolase 1
Gene Aliases
Aadacl1, RGD1311104
Chromosome Location
Chr.2: 112868834 - 112932622 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127524.2 NP_001120996.1 4 - 5 656 70
GenBank mRNA BC166533.1 - 4 - 5 656 70

View Full Product Details