The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037183.1 | NP_001032260.1 | 6 - 7 | 635 | 60 |
XM_008760912.1 | XP_008759134.1 | 5 - 6 | 472 | 60 | |
GenBank mRNA | BC089884.1 | - | 6 - 7 | 636 | 60 |
BC105820.1 | - | 6 - 7 | 635 | 60 | |
FQ222657.1 | - | 1 - 2 | 80 | 60 |