The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107668.1 | NP_001101138.1 | 4 - 5 | 665 | 140 |
XM_003749267.3 | XP_003749315.1 | - | 544 | 140 | |
XM_006232261.2 | XP_006232323.1 | 5 - 6 | 663 | 140 | |
XM_006232262.2 | XP_006232324.1 | 4 - 5 | 876 | 140 | |
XM_006232263.2 | XP_006232325.1 | 4 - 5 | 591 | 140 | |
XM_006232264.2 | XP_006232326.1 | 4 - 5 | 1331 | 140 | |
XM_008760925.1 | XP_008759147.1 | 4 - 5 | 650 | 140 | |
XM_008760926.1 | XP_008759148.1 | - | 682 | 140 | |
XM_008760927.1 | XP_008759149.1 | 4 - 5 | 2103 | 140 |