Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubl3
Entrez Gene ID
363869
Gene Name
ubiquitin-like 3
Gene Aliases
-
Chromosome Location
Chr.12: 7865938 - 7911526 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015030.1 NP_001015030.1 3 - 4 1012 105
GenBank mRNA BC091342.1 - 3 - 4 1012 105

View Full Product Details