Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr95
Entrez Gene ID
100361588
Gene Name
WD40 repeat domain 95
Gene Aliases
RGD1561029
Chromosome Location
Chr.12: 6265577 - 6317097 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769053.1 XP_008767275.1 8 - 9 883 65

View Full Product Details