Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc46a3
Entrez Gene ID
288454
Gene Name
solute carrier family 46, member 3
Gene Aliases
RGD1307594
Chromosome Location
Chr.12: 8725480 - 8741944 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024968.1 NP_001020139.1 5 - 6 1654 54
XM_006248833.2 XP_006248895.1 5 - 6 1482 54
GenBank mRNA BC091179.1 - 5 - 6 1654 54

View Full Product Details