The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022544.2 | NP_071989.1 | 1 - 2 | 193 | 105 |
GenBank mRNA | AY621374.1 | - | 1 - 2 | 193 | 105 |