Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plekha2
Entrez Gene ID
684785
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2
Gene Aliases
-
Chromosome Location
Chr.16: 71725659 - 71784432 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751621.3 XP_003751669.1 8 - 9 873 87

View Full Product Details