The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134454.1 | NP_604449.1 | 8 - 9 | 1403 | 67 |
GenBank mRNA | AF030378.1 | - | 2 - 3 | 315 | 67 |
AF311728.1 | - | 2 - 3 | 315 | 67 | |
BC161931.1 | - | 8 - 9 | 1403 | 67 |