The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191555.1 | NP_001178484.1 | 5 - 6 | 3395 | 80 |
NM_001197241.1 | NP_001184170.1 | 4 - 5 | 1772 | 80 | |
XM_008771331.1 | XP_008769553.1 | 5 - 6 | 3652 | 80 | |
GenBank mRNA | BC107469.1 | - | 4 - 5 | 1772 | 80 |
FQ221846.1 | - | 4 - 5 | 1781 | 80 |