Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Yipf3
Entrez Gene ID
301245
Gene Name
Yip1 domain family, member 3
Gene Aliases
C6orf109, KLIP1, LRRGT00110, RGD1308185
Chromosome Location
Chr.9: 17115308 - 17120618 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007801.2 NP_001007802.1 8 - 9 1001 76
GenBank mRNA AY387096.1 - 8 - 9 896 76
BC087102.1 - 8 - 9 1001 76

View Full Product Details