The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004224.1 | NP_001004224.1 | 7 - 8 | 788 | 69 |
XM_006229556.2 | XP_006229618.1 | 6 - 7 | 745 | 69 | |
XM_006229557.2 | XP_006229619.1 | 7 - 8 | 862 | 69 | |
GenBank mRNA | BC079182.1 | - | 7 - 8 | 788 | 69 |
FQ230132.1 | - | 7 - 8 | 779 | 69 | |
FQ230136.1 | - | 7 - 8 | 779 | 69 |