Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw17
Entrez Gene ID
361219
Gene Name
F-box and WD-40 domain protein 17
Gene Aliases
RGD1566133
Chromosome Location
Chr.17: 15076773 - 15094620 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082409.1 NP_001075878.1 1 - 2 105 99
XM_008771539.1 XP_008769761.1 1 - 2 150 99
XM_008771540.1 XP_008769762.1 1 - 2 150 99
XM_008771541.1 XP_008769763.1 1 - 2 150 99
GenBank mRNA BC167744.1 - 1 - 2 94 99

View Full Product Details