Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ror2
Entrez Gene ID
306782
Gene Name
receptor tyrosine kinase-like orphan receptor 2
Gene Aliases
-
Chromosome Location
Chr.17: 11953552 - 12134386 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107339.1 NP_001100809.1 2 - 3 369 57
XM_008771495.1 XP_008769717.1 1 - 2 81 57

View Full Product Details