The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001511.2 | NP_001001511.1 | 1 - 2 | 1400 | 91 |
GenBank mRNA | AY435153.1 | - | 1 - 2 | 920 | 91 |
BC099796.1 | - | 1 - 2 | 1400 | 91 |