Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gcnt2
Entrez Gene ID
306860
Gene Name
glucosaminyl (N-acetyl) transferase 2, I-branching enzyme
Gene Aliases
Ignt
Chromosome Location
Chr.17: 21634546 - 21677477 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001511.2 NP_001001511.1 2 - 3 1496 78
GenBank mRNA AY435151.1 - - 1016 78
AY435152.1 - - 1016 78
AY435153.1 - 2 - 3 1016 78
BC099796.1 - 2 - 3 1496 78

View Full Product Details