Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd34b
Entrez Gene ID
499506
Gene Name
ankyrin repeat domain 34B
Gene Aliases
Dp58
Chromosome Location
Chr.2: 22000106 - 22016030 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109174.1 NP_001102644.1 2 - 3 454 65

View Full Product Details